Product Details

SNP ID
rs13390810
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:69013206 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGAACTCCTTCCATTGCAAAAGCTC[A/G]GCGCGGCCTCGGGAGCTGCCCGGCG
Phenotype
MIM: 606410
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ANTXR1 PubMed Links

Gene Details

Gene
ANTXR1
Gene Name
anthrax toxin receptor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018153.3 63 UTR 5 NP_060623.2
NM_032208.2 63 UTR 5 NP_115584.1
NM_053034.2 63 UTR 5 NP_444262.1
XM_017005075.1 63 Intron XP_016860564.1
XM_017005076.1 63 Intron XP_016860565.1
XM_017005077.1 63 Intron XP_016860566.1

View Full Product Details