Product Details
- SNP ID
-
rs11126180
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:68136779 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TCTATTGGTCTAACGCCATAATACC[A/G]CATTTCCCTCTCAGCTACCTCAAAT
- Phenotype
-
MIM: 610729
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
WDR92
PubMed Links
Gene Details
- Gene
- WDR92
- Gene Name
- WD repeat domain 92
View Full Product Details