Product Details

SNP ID
rs13420146
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:8008817 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACAAAATCAAATCCAGGCCTGGTAG[C/T]TACCTATTTCATTAAGGAGCATTTA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LINC00299 PubMed Links

Gene Details

Gene
LINC00299
Gene Name
long intergenic non-protein coding RNA 299
There are no transcripts associated with this gene.

View Full Product Details