Product Details

SNP ID
rs7163110
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:88504107 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTCTCATGAGGTTGCTGTCATCTA[C/T]AGGCTTGAATGAAGCGGGAGGACTG
Phenotype
MIM: 608727
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DET1 PubMed Links

Gene Details

Gene
DET1
Gene Name
de-etiolated homolog 1 (Arabidopsis)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001144074.2 Intron NP_001137546.1
NM_001321594.1 Intron NP_001308523.1
NM_001321596.1 Intron NP_001308525.1
NM_017996.4 Intron NP_060466.2

View Full Product Details