Product Details

SNP ID
rs9555787
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:111311887 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CATTAATCTTAGGTGGATGTTAAAT[A/G]TAAATGTGCAGGCGAGGCCGAGCGT
Phenotype
MIM: 605477
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ARHGEF7 PubMed Links

Gene Details

Gene
ARHGEF7
Gene Name
Rho guanine nucleotide exchange factor 7
There are no transcripts associated with this gene.

Gene
TEX29
Gene Name
testis expressed 29
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001303133.1 Intron NP_001290062.1
NM_152324.2 Intron NP_689537.1
XM_017020387.1 Intron XP_016875876.1
XM_017020388.1 Intron XP_016875877.1

View Full Product Details