Product Details

SNP ID
rs13415695
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:190412298 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGGTAGTAGGTAATCTTGAGAAAG[A/G]GGGAATTGTAAAAGTATTTTACAGA
Phenotype
MIM: 613476
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MFSD6 PubMed Links

Gene Details

Gene
MFSD6
Gene Name
major facilitator superfamily domain containing 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017694.3 Intron NP_060164.3
XM_005246654.2 Intron XP_005246711.1
XM_005246656.3 Intron XP_005246713.1
XM_011511368.1 Intron XP_011509670.1
XM_017004392.1 Intron XP_016859881.1

View Full Product Details