Product Details
- SNP ID
-
rs8110240
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:48494890 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AGTGCCCTCTCTAGAACACTCTCCC[A/G]CCGATGTCCACCTGCCTCACTCCCT
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
LMTK3
PubMed Links
Gene Details
- Gene
- LMTK3
- Gene Name
- lemur tyrosine kinase 3
View Full Product Details