Product Details

SNP ID
rs12161539
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:160092316 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGGGTAGACTGTAACAGGCCCTGA[A/G]CGGGCACTGCCCGCCTGGTACCAGC
Phenotype
MIM: 606644 MIM: 600932
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
IGSF8 PubMed Links

Gene Details

Gene
IGSF8
Gene Name
immunoglobulin superfamily member 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001206665.2 1773 Silent Mutation CGC,CGT R564R NP_001193594.1
NM_001320247.1 1773 Silent Mutation CGC,CGT R564R NP_001307176.1
NM_052868.5 1773 Silent Mutation CGC,CGT R564R NP_443100.1
XM_006711631.3 1773 Silent Mutation CGC,CGT R547R XP_006711694.1
XM_011510159.2 1773 Silent Mutation CGC,CGT R542R XP_011508461.1
XM_017002835.1 1773 Silent Mutation CGC,CGT R564R XP_016858324.1
XM_017002836.1 1773 Silent Mutation CGC,CGT R564R XP_016858325.1
XM_017002837.1 1773 Silent Mutation CGC,CGT R564R XP_016858326.1
XM_017002838.1 1773 Silent Mutation CGC,CGT R542R XP_016858327.1
XM_017002839.1 1773 Silent Mutation CGC,CGT R542R XP_016858328.1
XM_017002840.1 1773 Silent Mutation CGC,CGT R542R XP_016858329.1
Gene
KCNJ9
Gene Name
potassium voltage-gated channel subfamily J member 9
There are no transcripts associated with this gene.

View Full Product Details