Product Details

SNP ID
rs10981225
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:112043701 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAAATGCTGCCAAGAGCCTTTCTAG[A/G]AAACATGGCTTATATTATTTGGTTG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SUSD1 PubMed Links

Gene Details

Gene
SUSD1
Gene Name
sushi domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282640.1 Intron NP_001269569.1
NM_001282643.1 Intron NP_001269572.1
NM_022486.4 Intron NP_071931.2
XM_011518915.1 Intron XP_011517217.1
XM_011518916.2 Intron XP_011517218.1
XM_011518917.1 Intron XP_011517219.1
XM_011518918.1 Intron XP_011517220.1
XM_011518919.1 Intron XP_011517221.1
XM_017015037.1 Intron XP_016870526.1
XM_017015038.1 Intron XP_016870527.1
XM_017015039.1 Intron XP_016870528.1
XM_017015040.1 Intron XP_016870529.1

View Full Product Details