Product Details

SNP ID
rs11796239
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:47447154 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTACTATTTCCAGGAGCTCAAGAG[G/T]TCTGTATGAATCATGTCACCAAACA
Phenotype
MIM: 314995
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ZNF41 PubMed Links

Gene Details

Gene
ZNF41
Gene Name
zinc finger protein 41
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001324139.1 3383 UTR 3 NP_001311068.1
NM_001324140.1 3383 UTR 3 NP_001311069.1
NM_001324141.1 3383 UTR 3 NP_001311070.1
NM_001324142.1 3383 UTR 3 NP_001311071.1
NM_001324143.1 3383 UTR 3 NP_001311072.1
NM_001324144.1 3383 UTR 3 NP_001311073.1
NM_001324145.1 3383 UTR 3 NP_001311074.1
NM_001324147.1 3383 UTR 3 NP_001311076.1
NM_001324148.1 3383 UTR 3 NP_001311077.1
NM_001324149.1 3383 UTR 3 NP_001311078.1
NM_001324150.1 3383 UTR 3 NP_001311079.1
NM_001324151.1 3383 UTR 3 NP_001311080.1
NM_001324152.1 3383 UTR 3 NP_001311081.1
NM_001324153.1 3383 UTR 3 NP_001311082.1
NM_001324154.1 3383 UTR 3 NP_001311083.1
NM_001324155.1 3383 UTR 3 NP_001311084.1
NM_001324156.1 3383 UTR 3 NP_001311085.1
NM_001324157.1 3383 UTR 3 NP_001311086.1
NM_007130.3 3383 UTR 3 NP_009061.1
NM_153380.3 3383 UTR 3 NP_700359.1
XM_006724550.3 3383 UTR 3 XP_006724613.1
XM_006724555.3 3383 UTR 3 XP_006724618.1
XM_017029810.1 3383 UTR 3 XP_016885299.1
XM_017029811.1 3383 UTR 3 XP_016885300.1
XM_017029812.1 3383 UTR 3 XP_016885301.1
XM_017029813.1 3383 UTR 3 XP_016885302.1
XM_017029814.1 3383 UTR 3 XP_016885303.1
XM_017029815.1 3383 UTR 3 XP_016885304.1
XM_017029816.1 3383 UTR 3 XP_016885305.1
XM_017029817.1 3383 UTR 3 XP_016885306.1

View Full Product Details