Product Details

SNP ID
rs11090091
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:42381656 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCACTCAGGACTGCTCACATCCGTC[C/T]GTCCTGGAAGGTCCACAGCTCCCCT
Phenotype
MIM: 608740
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NFAM1 PubMed Links

Gene Details

Gene
NFAM1
Gene Name
NFAT activating protein with ITAM motif 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318323.1 4983 UTR 3 NP_001305252.1
NM_145912.6 4983 UTR 3 NP_666017.1
XM_005261374.4 4983 UTR 3 XP_005261431.1

View Full Product Details