Product Details

SNP ID
rs7906870
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:92296310 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAAGGGGGGTGATTTTTCTAGTGTC[A/G]TGAATGTTTCAGTGTCACAAATATC
Phenotype
MIM: 610606 MIM: 610637
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CPEB3 PubMed Links

Gene Details

Gene
CPEB3
Gene Name
cytoplasmic polyadenylation element binding protein 3
There are no transcripts associated with this gene.

Gene
MARCH5
Gene Name
membrane associated ring-CH-type finger 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017824.4 Intron NP_060294.1
XM_011539886.2 Intron XP_011538188.1

View Full Product Details