Product Details
- SNP ID
-
rs12355840
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:133247608 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATGCATAGGAAAAAGGAACGGCGGG[C/T]GGCTCTGAGGCGGGCCGGCTCTTCA
- Phenotype
-
MIM: 607158
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
MIR202
PubMed Links
Gene Details
- Gene
- MIR202
- Gene Name
- microRNA 202
There are no transcripts associated with this gene.
- Gene
- MIR202HG
- Gene Name
- MIR202 host gene
There are no transcripts associated with this gene.
- Gene
- VENTX
- Gene Name
- VENT homeobox
There are no transcripts associated with this gene.
View Full Product Details