Product Details

SNP ID
rs4737066
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:42708032 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTCAGGGACGGAGAACAGCTTGGGG[A/G]CAGGGCCCATCTCTGTGAAGAGGGA
Phenotype
MIM: 118508
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CHRNB3 PubMed Links

Gene Details

Gene
CHRNB3
Gene Name
cholinergic receptor nicotinic beta 3 subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000749.3 Intron NP_000740.1
XM_011544390.1 Intron XP_011542692.1

View Full Product Details