Product Details

SNP ID
rs11624298
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:23383943 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATCTATCAAATGGACTGACACCCTC[A/G]TCTCCTTGGCCTTTTCTTCCCAGGT
Phenotype
MIM: 607888 MIM: 605658 MIM: 611116 MIM: 160710
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CMTM5 PubMed Links

Gene Details

Gene
CMTM5
Gene Name
CKLF like MARVEL transmembrane domain containing 5
There are no transcripts associated with this gene.

Gene
IL25
Gene Name
interleukin 25
There are no transcripts associated with this gene.

Gene
MIR208A
Gene Name
microRNA 208a
There are no transcripts associated with this gene.

Gene
MYH6
Gene Name
myosin heavy chain 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002471.3 Intron NP_002462.2

View Full Product Details