Product Details

SNP ID
rs12279755
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:111702629 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAGCACTTAGCACAGCGCCTGCCAT[A/G]TAGTGACAGGAGGTTACCTTATGTG
Phenotype
MIM: 603113 MIM: 608973
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PPP2R1B PubMed Links

Gene Details

Gene
PPP2R1B
Gene Name
protein phosphatase 2 scaffold subunit Abeta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001177562.1 Intron NP_001171033.1
NM_001177563.1 Intron NP_001171034.1
NM_002716.4 Intron NP_002707.3
NM_181699.2 Intron NP_859050.1
NM_181700.1 Intron NP_859051.1
XM_017017958.1 Intron XP_016873447.1
XM_017017959.1 Intron XP_016873448.1
XM_017017960.1 Intron XP_016873449.1
XM_017017961.1 Intron XP_016873450.1
Gene
SIK2
Gene Name
salt inducible kinase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015191.2 Intron NP_056006.1
XM_017017417.1 Intron XP_016872906.1
XM_017017418.1 Intron XP_016872907.1

View Full Product Details