Product Details
- SNP ID
-
rs11601189
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:122977765 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGGCTCAGCCGAGCCTCGGCGGCGG[G/T]GGCGGCCTCTGAACCGCGGGGGCTG
- Phenotype
-
MIM: 611074
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
BSX
PubMed Links
Gene Details
- Gene
- BSX
- Gene Name
- brain specific homeobox
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001098169.1 |
586 |
Missense Mutation |
ACC,CCC |
T196P |
NP_001091639.1 |
View Full Product Details