Product Details
- SNP ID
-
rs7103769
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:134233936 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGGCATGAACTTATCATCAAAAAAT[A/T]CCAGAACTATTTCCTGAAGAGGATA
- Phenotype
-
MIM: 609276
MIM: 610027
- Polymorphism
- A/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
NCAPD3
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs113700919] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- NCAPD3
- Gene Name
- non-SMC condensin II complex subunit D3
There are no transcripts associated with this gene.
- Gene
- VPS26B
- Gene Name
- VPS26, retromer complex component B
View Full Product Details