Product Details

SNP ID
rs13412814
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:217284485 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTAAGTTTATATAATCAGTACTTC[A/G]TAATTCAGAAACTTTGCCTCTCAGA
Phenotype
MIM: 608262
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DIRC3 PubMed Links

Gene Details

Gene
DIRC3
Gene Name
disrupted in renal carcinoma 3
There are no transcripts associated with this gene.

Gene
DIRC3-AS1
Gene Name
DIRC3 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details