Product Details

SNP ID
rs13397731
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:230330367 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCTTCTCTGCCCCTGGCCTGTAAGA[G/T]CCTCTTCTTATGCCAGTACCACACT
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
SP140L PubMed Links

Gene Details

Gene
SP140L
Gene Name
SP140 nuclear body protein like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001308162.1 Intron NP_001295091.1
NM_001308163.1 Intron NP_001295092.1
NM_138402.4 Intron NP_612411.4
XM_006712855.2 Intron XP_006712918.1
XM_006712856.2 Intron XP_006712919.1
XM_006712858.3 Intron XP_006712921.1
XM_011512190.2 Intron XP_011510492.1
XM_011512191.1 Intron XP_011510493.1
XM_017005294.1 Intron XP_016860783.1
XM_017005295.1 Intron XP_016860784.1
XM_017005296.1 Intron XP_016860785.1
XM_017005297.1 Intron XP_016860786.1
XM_017005298.1 Intron XP_016860787.1
XM_017005299.1 Intron XP_016860788.1
XM_017005300.1 Intron XP_016860789.1
XM_017005301.1 Intron XP_016860790.1

View Full Product Details