Product Details

SNP ID
rs9657994
Assay Type
Functionally tested
NCBI dbSNP Submissions
5
Location
Chr.1:7931641 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTAGATGTGGTTAGCAGTAGTGGGC[A/G]AAGAGGGCTGTGATCTAGAAAGGGC
Phenotype
MIM: 602250
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TNFRSF9 PubMed Links

Gene Details

Gene
TNFRSF9
Gene Name
TNF receptor superfamily member 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001561.5 Intron NP_001552.2
XM_006710618.3 Intron XP_006710681.1
XM_011541386.2 Intron XP_011539688.1

View Full Product Details