Product Details

SNP ID
rs11586116
Assay Type
Functionally Tested
NCBI dbSNP Submissions
3
Location
Chr.1:8013496 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACCTTACTGGCAGATCCTTCGCTG[A/T]TCTGTCTTTGCAGTGCTTTGCTGCT
Phenotype
MIM: 608069
Polymorphism
A/T, Transversion Substitution
Allele Nomenclature
Literature Links
ERRFI1 PubMed Links

Gene Details

Gene
ERRFI1
Gene Name
ERBB receptor feedback inhibitor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018948.3 1366 Missense Mutation AAC,ATC N368I NP_061821.1
XM_005263477.2 1366 Missense Mutation AAC,ATC N317I XP_005263534.1
XM_006710697.2 1366 Missense Mutation AAC,ATC N368I XP_006710760.1
XM_011541596.2 1366 Missense Mutation AAC,ATC N293I XP_011539898.1

View Full Product Details