Product Details

SNP ID
rs12161033
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:43530270 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGACTGAGATTTATACTTAACCCC[A/G]AAGGAAAGAGGAACGTGAATGTGGA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
EFCAB6 PubMed Links

Gene Details

Gene
EFCAB6
Gene Name
EF-hand calcium binding domain 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022785.3 Intron NP_073622.2
NM_198856.2 Intron NP_942153.1
XM_005261704.2 Intron XP_005261761.1
XM_005261705.3 Intron XP_005261762.1
XM_011530316.1 Intron XP_011528618.1
XM_011530317.2 Intron XP_011528619.1
XM_011530318.2 Intron XP_011528620.1
XM_011530319.2 Intron XP_011528621.1
XM_011530320.2 Intron XP_011528622.1
XM_011530321.2 Intron XP_011528623.1
XM_011530322.2 Intron XP_011528624.1
XM_011530323.2 Intron XP_011528625.1
XM_011530325.2 Intron XP_011528627.1
XM_011530326.2 Intron XP_011528628.1
XM_011530327.2 Intron XP_011528629.1
XM_011530328.2 Intron XP_011528630.1
XM_011530329.1 Intron XP_011528631.1
XM_011530330.2 Intron XP_011528632.1
XM_017028910.1 Intron XP_016884399.1
XM_017028911.1 Intron XP_016884400.1
XM_017028912.1 Intron XP_016884401.1
Gene
EFCAB6-AS1
Gene Name
EFCAB6 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details