Product Details

SNP ID
rs11230188
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:60204188 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAGAATCAGGAGATGTAAGGACTTC[C/T]AGGATTTAGTATGAGTTCTGGCTTT
Phenotype
MIM: 608401
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MIR6503 PubMed Links

Gene Details

Gene
MIR6503
Gene Name
microRNA 6503
There are no transcripts associated with this gene.

Gene
MS4A4E
Gene Name
membrane spanning 4-domains A4E
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_011545416.2 1109 UTR 3 XP_011543718.1
XM_011545417.2 1109 Intron XP_011543719.1
XM_011545418.2 1109 Intron XP_011543720.1
XM_011545419.2 1109 Intron XP_011543721.1
XM_017018641.1 1109 Intron XP_016874130.1

View Full Product Details