Product Details

SNP ID
rs11906246
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:33818848 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTTCATTGACTGTCCATTTTTTGG[C/T]GCTTCTGTTCCTTCCTTTTTACTCT
Phenotype
MIM: 610897
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CHMP4B PubMed Links

Gene Details

Gene
CHMP4B
Gene Name
charged multivesicular body protein 4B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_176812.4 Intron NP_789782.1
Gene
ZNF341-AS1
Gene Name
ZNF341 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details