Product Details
- SNP ID
-
rs7621631
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:169794357 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- ATCTACAGATTTAGGCTAAAACAGA[A/C]AATTCCATAGTATTGGATGCGGACC
- Phenotype
-
MIM: 606042
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
LRRC34
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs7643115] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- LRRC34
- Gene Name
- leucine rich repeat containing 34
- Gene
- MYNN
- Gene Name
- myoneurin
There are no transcripts associated with this gene.
View Full Product Details