Product Details

SNP ID
hCV32105843
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:721846 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGGCCTCCGCCCGGCCGTGGGCTAC[G/T]AGCTGAACCCCTGGCTGGTGGCGCT
Phenotype
MIM: 614666 MIM: 610998 MIM: 611118
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CCDC78 PubMed Links

Gene Details

Gene
CCDC78
Gene Name
coiled-coil domain containing 78
There are no transcripts associated with this gene.

Gene
FAM173A
Gene Name
family with sequence similarity 173 member A
There are no transcripts associated with this gene.

Gene
HAGHL
Gene Name
hydroxyacylglutathione hydrolase-like
There are no transcripts associated with this gene.

Gene
METRN
Gene Name
meteorin, glial cell differentiation regulator
There are no transcripts associated with this gene.

Gene
NARFL
Gene Name
nuclear prelamin A recognition factor like
There are no transcripts associated with this gene.

View Full Product Details