Product Details

SNP ID
rs10733604
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:113279983 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGTCTCTCTGTCCACTGTCTCATCT[A/C]CCCTTCATCATCCAGCTTGTTGGAA
Phenotype
MIM: 614533 MIM: 607795
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CDC26 PubMed Links

Gene Details

Gene
CDC26
Gene Name
cell division cycle 26
There are no transcripts associated with this gene.

Gene
PRPF4
Gene Name
pre-mRNA processing factor 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001244926.1 Intron NP_001231855.1
NM_001322266.1 Intron NP_001309195.1
NM_001322267.1 Intron NP_001309196.1
NM_004697.4 Intron NP_004688.2

View Full Product Details