Product Details

SNP ID
rs8177817
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.9:113388943 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCAGAGTGCTTATCAGTCCCTGTGG[C/T]GCAGGTCAAAACACCCCACCCTTGC
Phenotype
MIM: 125270
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
ALAD PubMed Links

Gene Details

Gene
ALAD
Gene Name
aminolevulinate dehydratase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000031.5 Intron NP_000022.3
NM_001003945.2 Intron NP_001003945.1
NM_001317745.1 Intron NP_001304674.1
XM_011518364.2 Intron XP_011516666.1

View Full Product Details