Product Details

SNP ID
rs9341303
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.Y:12913594 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTGTAAACTAAATCAGACAGTTTA[A/G]TTGGTTACTTCCATTAATATGTTAG
Phenotype
MIM: 400010
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DDX3Y PubMed Links

Gene Details

Gene
DDX3Y
Gene Name
DEAD-box helicase 3, Y-linked
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001122665.3 Intron NP_001116137.1
NM_001302552.2 Intron NP_001289481.1
NM_001324195.1 Intron NP_001311124.1
NM_004660.4 Intron NP_004651.2

View Full Product Details