Product Details

SNP ID
rs12542891
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:100257276 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTTTCTTTTTACCAAAAGAAAACG[C/T]TCAGTAGCACCATAATGGTAATACT
Phenotype
MIM: 607119
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RNF19A PubMed Links

Gene Details

Gene
RNF19A
Gene Name
ring finger protein 19A, RBR E3 ubiquitin protein ligase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001280539.1 3970 UTR 3 NP_001267468.1
NM_015435.4 3970 UTR 3 NP_056250.3
NM_183419.3 3970 UTR 3 NP_904355.1
XM_005250853.3 3970 Intron XP_005250910.1
XM_011516956.2 3970 UTR 3 XP_011515258.1
XM_017013302.1 3970 Intron XP_016868791.1
XM_017013303.1 3970 Intron XP_016868792.1
XM_017013304.1 3970 UTR 3 XP_016868793.1
XM_017013305.1 3970 UTR 3 XP_016868794.1

View Full Product Details