Product Details

SNP ID
rs12094370
Assay Type
Functionally tested
NCBI dbSNP Submissions
35
Location
Chr.1:219910817 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GATCATCTAGCAAACATTCAAGAAA[A/G]TTCCATGTAAACAAAAAGGTACGAA
Phenotype
MIM: 611146
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SLC30A10 PubMed Links

Gene Details

Gene
SLC30A10
Gene Name
solute carrier family 30 member 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018713.2 6637 Intron NP_061183.2
XM_006711437.3 6637 UTR 3 XP_006711500.2
XM_017001684.1 6637 UTR 3 XP_016857173.1

View Full Product Details