Product Details

SNP ID
rs11052384
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:9681885 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AATATACTTTATACATTTATGAAGC[A/C]AAAATAATTTTTTTATTTCAGAGAT
Phenotype
MIM: 605659
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CLEC2D PubMed Links

Gene Details

Gene
CLEC2D
Gene Name
C-type lectin domain family 2 member D
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004419.4 Intron NP_001004419.1
NM_001197317.2 Intron NP_001184246.1
NM_001197318.2 Intron NP_001184247.1
NM_001197319.2 Intron NP_001184248.1
NM_013269.5 Intron NP_037401.1

View Full Product Details