Product Details
- SNP ID
-
rs11053565
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:10040253 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GAACTCAGCTGAGAATCCTGTATTC[A/T]AGAAAATGTCAACAACCACTTTGGG
- Phenotype
-
MIM: 612252
- Polymorphism
- A/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
CLEC9A
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs78064987] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CLEC9A
- Gene Name
- C-type lectin domain family 9 member A
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_207345.3 |
|
Intron |
|
|
NP_997228.1 |
- Gene
- LOC102724020
- Gene Name
- uncharacterized LOC102724020
There are no transcripts associated with this gene.
View Full Product Details