Product Details

SNP ID
rs11053566
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:10042166 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TATTCATGCAATTTTTTTGAGAAAA[G/T]AGAAAGGTATAGTTGAATTTCAAGT
Phenotype
MIM: 612252
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CLEC9A PubMed Links

Gene Details

Gene
CLEC9A
Gene Name
C-type lectin domain family 9 member A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_207345.3 Intron NP_997228.1

View Full Product Details