Product Details

SNP ID
rs13026227
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:54732966 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCAGCTGAGGAGGTGAGTGGTATG[A/G]AAAGCAGGGGGAGTTGTAACAGAGG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
EML6 PubMed Links

Gene Details

Gene
EML6
Gene Name
echinoderm microtubule associated protein like 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001039753.2 Intron NP_001034842.2
XM_017004098.1 Intron XP_016859587.1
XM_017004099.1 Intron XP_016859588.1
XM_017004100.1 Intron XP_016859589.1
XM_017004101.1 Intron XP_016859590.1
XM_017004102.1 Intron XP_016859591.1

View Full Product Details