Product Details
- SNP ID
-
rs11571911
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:44390288 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TTTAGGATGAAATTCTGAAAGCAGC[A/G]GTAATGAAATATGGGAAAAATCAGT
- Phenotype
-
MIM: 602868
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CDC5L
PubMed Links
Gene Details
- Gene
- CDC5L
- Gene Name
- cell division cycle 5 like
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001253.3 |
376 |
Silent Mutation |
GCA,GCG |
A22A |
NP_001244.1 |
View Full Product Details