Product Details

SNP ID
rs12697992
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:156675134 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TAGTGCAGACCCGAAGAGGCAGCGG[A/C]GACCCGAAGGGGCAGCGGAGACCCG
Phenotype
MIM: 605522 MIM: 610241
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
LMBR1 PubMed Links

Gene Details

Gene
LMBR1
Gene Name
limb development membrane protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022458.3 Intron NP_071903.2
XM_005249555.2 Intron XP_005249612.1
XM_005249558.2 Intron XP_005249615.1
XM_017012507.1 Intron XP_016867996.1
XM_017012508.1 Intron XP_016867997.1
XM_017012509.1 Intron XP_016867998.1
XM_017012510.1 Intron XP_016867999.1
XM_017012511.1 Intron XP_016868000.1
XM_017012512.1 Intron XP_016868001.1
XM_017012513.1 Intron XP_016868002.1
XM_017012514.1 Intron XP_016868003.1
XM_017012515.1 Intron XP_016868004.1
XM_017012516.1 Intron XP_016868005.1
XM_017012517.1 Intron XP_016868006.1
XM_017012518.1 Intron XP_016868007.1
Gene
RNF32
Gene Name
ring finger protein 32
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001184996.1 Intron NP_001171925.1
NM_001184997.1 Intron NP_001171926.1
NM_001308273.1 Intron NP_001295202.1
NM_001308274.1 Intron NP_001295203.1
NM_030936.3 Intron NP_112198.1
XM_005249522.4 Intron XP_005249579.1
XM_011515804.2 Intron XP_011514106.1
XM_011515805.2 Intron XP_011514107.1
XM_011515806.2 Intron XP_011514108.1
XM_011515807.2 Intron XP_011514109.1
XM_011515808.2 Intron XP_011514110.1
XM_011515809.2 Intron XP_011514111.1
XM_011515810.2 Intron XP_011514112.1
XM_011515811.2 Intron XP_011514113.1
XM_011515812.2 Intron XP_011514114.1
XM_011515813.2 Intron XP_011514115.1
XM_017011753.1 Intron XP_016867242.1
XM_017011754.1 Intron XP_016867243.1
XM_017011755.1 Intron XP_016867244.1

View Full Product Details