Product Details

SNP ID
rs13140929
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:107837664 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATTTACATCTGTAATAGATCCCTC[G/T]GGCTGCTGGGCAGAGAGTGGAGTAG
Phenotype
MIM: 611574
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
SGMS2 PubMed Links

Gene Details

Gene
SGMS2
Gene Name
sphingomyelin synthase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001136257.1 Intron NP_001129729.1
NM_001136258.1 Intron NP_001129730.1
NM_152621.5 Intron NP_689834.1
XM_011531698.2 Intron XP_011530000.1
XM_011531699.2 Intron XP_011530001.1
XM_011531700.2 Intron XP_011530002.1
XM_011531701.2 Intron XP_011530003.1
XM_011531702.2 Intron XP_011530004.1
XM_017007839.1 Intron XP_016863328.1
XM_017007840.1 Intron XP_016863329.1
XM_017007841.1 Intron XP_016863330.1

View Full Product Details