Product Details

SNP ID
rs12027405
Assay Type
Functionally tested
NCBI dbSNP Submissions
3
Location
Chr.1:67704245 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCTTGGGTAGCCTATGTGCCTCTT[A/G]GATGGTATGTGAACAAGGTAGGGAT
Phenotype
MIM: 615405
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GNG12 PubMed Links

Gene Details

Gene
GNG12
Gene Name
G protein subunit gamma 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018841.5 1630 UTR 3 NP_061329.3
XM_017001809.1 1630 UTR 3 XP_016857298.1
XM_017001810.1 1630 UTR 3 XP_016857299.1
XM_017001811.1 1630 UTR 3 XP_016857300.1

View Full Product Details