Product Details

SNP ID
rs13437457
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:136924186 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAACTTCACCACGCTGCACTGCAC[C/G]TACATCAATTCGGCCGTGACCACCA
Phenotype
MIM: 612519
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
SLC35D3 PubMed Links

Gene Details

Gene
SLC35D3
Gene Name
solute carrier family 35 member D3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001008783.2 929 Silent Mutation ACC,ACG T247T NP_001008783.1

View Full Product Details