Product Details

SNP ID
rs7275802
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:32785508 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCTGTAATTGATCGTTTTGATGTTC[A/T]TATTGTCCCTAATTTGGCCAGTAGA
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
C21orf62 PubMed Links

Gene Details

Gene
C21orf62
Gene Name
chromosome 21 open reading frame 62
There are no transcripts associated with this gene.

Gene
C21orf62-AS1
Gene Name
C21orf62 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details