Product Details

SNP ID
rs4916224
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:173610846 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAAGATATCACAAAATCTTAAAAAA[A/T]AAAAAATAATTCAGGTCTTAATTAT
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
ANKRD45 PubMed Links

Gene Details

Gene
ANKRD45
Gene Name
ankyrin repeat domain 45
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_198493.2 Intron NP_940895.1
Gene
LOC730159
Gene Name
uncharacterized LOC730159
There are no transcripts associated with this gene.

Gene
SLC9C2
Gene Name
solute carrier family 9 member C2 (putative)
There are no transcripts associated with this gene.

View Full Product Details