Product Details

SNP ID
rs4869248
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:95721800 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAAAAACAGAAAGTGAAAAAATGT[C/G]TGTAAGATAAGCCAAAATTGTTAAA
Phenotype
MIM: 607353 MIM: 608039
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
RHOBTB3 PubMed Links

Gene Details

Gene
RHOBTB3
Gene Name
Rho related BTB domain containing 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014899.3 Intron NP_055714.3
Gene
SPATA9
Gene Name
spermatogenesis associated 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_031952.3 Intron NP_114158.2

View Full Product Details