Product Details

SNP ID
rs4424506
Assay Type
Functionally tested
NCBI dbSNP Submissions
7
Location
Chr.1:112685775 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAATATAATAAGTTTGTGATTTTT[A/T]AAAAATAATTTTTGTATGACTGTTA
Phenotype
MIM: 610742
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
MOV10 PubMed Links

Gene Details

Gene
MOV10
Gene Name
Mov10 RISC complex RNA helicase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001130079.2 Intron NP_001123551.1
NM_001286072.1 Intron NP_001273001.1
NM_001321324.1 Intron NP_001308253.1
NM_020963.4 Intron NP_066014.1
XM_005270869.4 Intron XP_005270926.1
XM_005270870.4 Intron XP_005270927.1
XM_017001318.1 Intron XP_016856807.1
XM_017001319.1 Intron XP_016856808.1

View Full Product Details