Product Details

SNP ID
rs4805135
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:35651901 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTTTTTTTAAGCTGTATGTGTGTGC[A/G]TACTGTAACATGTACAGAAAGCACA
Phenotype
MIM: 124089 MIM: 609358
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
COX6B1 PubMed Links

Gene Details

Gene
COX6B1
Gene Name
cytochrome c oxidase subunit 6B1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001863.4 Intron NP_001854.1
Gene
ETV2
Gene Name
ETS variant 2
There are no transcripts associated with this gene.

View Full Product Details