Product Details
- SNP ID
-
rs4645743
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.22:35402054 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CTGCGAGGCTGGGGAACAGCCTGTG[C/T]GTTGGTCAGTGGTCCCCAAATATGT
- Phenotype
-
MIM: 141250
MIM: 602696
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
HMOX1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs737777] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- HMOX1
- Gene Name
- heme oxygenase 1
There are no transcripts associated with this gene.
- Gene
- MCM5
- Gene Name
- minichromosome maintenance complex component 5
View Full Product Details