Product Details

SNP ID
rs4252600
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.17:39700729 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGACTGGCCTGGGCTGGATTCCTTG[C/G]GGGGGTCCTCTGCCCTGCCCCTCCT
Phenotype
MIM: 164870
Polymorphism
C/G, Transversion Substitution
Allele Nomenclature
Literature Links
ERBB2 PubMed Links

Gene Details

Gene
ERBB2
Gene Name
erb-b2 receptor tyrosine kinase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005862.2 Intron NP_001005862.1
NM_001289936.1 Intron NP_001276865.1
NM_001289937.1 Intron NP_001276866.1
NM_001289938.1 Intron NP_001276867.1
NM_004448.3 Intron NP_004439.2

View Full Product Details