Product Details

SNP ID
rs4543035
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:48327911 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATAAGCATGACTTTTGCTAAAACCC[A/G]CCATTAACCAGAATAATATCTGAGT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SPINK8 PubMed Links

Gene Details

Gene
SPINK8
Gene Name
serine peptidase inhibitor, Kazal type 8 (putative)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080525.1 Intron NP_001073994.1
XM_017007046.1 Intron XP_016862535.1
XM_017007047.1 Intron XP_016862536.1

View Full Product Details