Product Details

SNP ID
rs4558175
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:119001871 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAGGAGTTTGAGGCCATCCTGGGCA[A/G]TGTATCAAGACCCCATCTCTAAAAA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CCDC84 PubMed Links

Gene Details

Gene
CCDC84
Gene Name
coiled-coil domain containing 84
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_198489.2 Intron NP_940891.1
XM_011542799.1 Intron XP_011541101.1
XM_011542800.2 Intron XP_011541102.1
XM_017017652.1 Intron XP_016873141.1
Gene
RPL23AP64
Gene Name
ribosomal protein L23a pseudogene 64
There are no transcripts associated with this gene.

View Full Product Details